Assay Details
Target Gene Details
Entrez Gene ID: | 347736 |
Gene Name: | NME/NM23 family member 9 |
Gene Aliases: |
NM23-H9, TXL-2, TXL2, TXNDC6 |
Location: |
Chr.3:138261437-138331785 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME9 | NM_178130.3 | 10 | 764 | NP_835231.1 |
| XM_005247429.3 | 10 | 745 | XP_005247486.1 | |
| XM_006713615.3 | 9 | 2627 | XP_006713678.1 | |
| XM_011512774.2 | 9 | 631 | XP_011511076.1 | |
| XM_011512777.2 | XP_011511079.1 | |||
| XM_017006306.1 | 10 | 748 | XP_016861795.1 | |
| XM_017006307.1 | 9 | 1197 | XP_016861796.1 | |
| XM_017006308.1 | 10 | 2744 | XP_016861797.1 | |
| XM_017006309.1 | 10 | 776 | XP_016861798.1 | |
| XM_017006310.1 | 9 | 706 | XP_016861799.1 | |
| XM_017006311.1 | 9 | 631 | XP_016861800.1 | |
| XM_017006312.1 | 9 | 2627 | XP_016861801.1 | |
| XM_017006313.1 | 11 | 3251 | XP_016861802.1 | |
| XM_017006314.1 | 10 | 1202 | XP_016861803.1 | |
| XM_017006315.1 | 8 | 680 | XP_016861804.1 | |
| XM_017006316.1 | 8 | 2555 | XP_016861805.1 | |
| XM_017006317.1 | 8 | 680 | XP_016861806.1 | |
| XM_017006318.1 | 7 | 1008 | XP_016861807.1 | |
| XM_017006319.1 | 5 | 415 | XP_016861808.1 | |
| AF196568.1 | 9 | 737 | AAG28497.1 | |
| AK094830.1 | 10 | 764 | BAC04430.1 | |
| AY336746.1 | 6 | 454 | AAQ16116.1 | |
| BC107054.2 | AAI07055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map