Assay Details
Target Gene Details
Entrez Gene ID: | 222546 |
Gene Name: | regulatory factor X6 |
Gene Aliases: |
MTCHRS, MTFS, RFXDC1, dJ955L16.1 |
Location: |
Chr.6:116877213-116932163 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX6 | NM_173560.3 | 1 | 198 | NP_775831.2 |
| XM_011535589.1 | 1 | 193 | XP_011533891.1 | |
| BC039248.1 | 1 | 198 | AAH39248.1 | |
| CA842450.1 | 1 | 129 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830784 | Chr.6:116845788 - 116984209 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map