Assay Details
Target Gene Details
Entrez Gene ID: | 339751 |
Gene Name: | MLK7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:173197712-173282036 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLK7-AS1 | NR_033882.1 |
Target Gene Details
Entrez Gene ID: | 51776 |
Gene Name: | sterile alpha motif and leucine zipper containing kinase AZK |
Gene Aliases: |
AZK, MLK7, MLT, MLTK, MRK, SFMMP, mlklak, pk |
Location: |
Chr.2:173075435-173268009 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZAK | NM_016653.2 | NP_057737.2 | ||
| NM_133646.2 | 12 | 2683 | NP_598407.1 | |
| XM_005246640.2 | XP_005246697.1 | |||
| XM_017004323.1 | 12 | 2729 | XP_016859812.1 | |
| XM_017004324.1 | 12 | 2753 | XP_016859813.1 | |
| AB030034.1 | BAB12040.1 | |||
| AB049733.1 | BAB16444.1 | |||
| AB208974.1 | BAD92211.1 | |||
| AF238255.1 | AAF63490.1 | |||
| AF251441.1 | AAF65822.1 | |||
| AF480461.1 | AAL85891.1 | |||
| AF480462.1 | 12 | 2678 | AAL85892.1 | |
| AL833180.1 | 3 | 1686 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3037 | Chr.2:173190146 - 173223386 on Build GRCh38 | Insertion |
|
| esv3584178 | Chr.2:173210824 - 173224533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map