Assay Details
Target Gene Details
Entrez Gene ID: | 7054 |
Gene Name: | tyrosine hydroxylase |
Gene Aliases: |
DYT14, DYT5b, TYH |
Location: |
Chr.11:2163929-2174081 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TH | NM_000360.3 | NP_000351.2 | ||
| NM_199292.2 | NP_954986.2 | |||
| NM_199293.2 | NP_954987.2 | |||
| XM_011520335.2 | XP_011518637.1 | |||
| BC104967.1 | AAI04968.1 | |||
| BC143611.1 | ||||
| BC143614.1 | ||||
| DQ677336.1 | ||||
| DQ677337.1 | ||||
| M17589.1 | AAA61179.1 | |||
| X05290.1 | CAA28908.1 | |||
| Y00414.1 | CAA68472.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553070 | Chr.11:2154812 - 2177188 on Build GRCh38 | Loss |
|
| nsv469927 | Chr.11:2165105 - 2201683 on Build GRCh38 | Loss |
|
| nsv553068 | Chr.11:2096173 - 2174751 on Build GRCh38 | Loss |
|
| nsv951284 | Chr.11:2083671 - 2218670 on Build GRCh38 | Deletion |
|
| nsv553047 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76240471] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map