Assay Details
Target Gene Details
Entrez Gene ID: | 10333 |
Gene Name: | toll like receptor 6 |
Gene Aliases: |
CD286 |
Location: |
Chr.4:38822897-38857767 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLR6 | NM_006068.4 | 2 | 920 | NP_006059.2 |
| XM_005262637.4 | 2 | 905 | XP_005262694.1 | |
| XM_011513612.2 | 3 | 1073 | XP_011511914.1 | |
| XM_011513613.2 | 2 | 1306 | XP_011511915.1 | |
| XM_011513614.2 | 2 | 943 | XP_011511916.1 | |
| AB020807.1 | 1 | 866 | BAA78631.1 | |
| AB445652.1 | 1 | 799 | ||
| AK313618.1 | 2 | 920 | ||
| BC111755.1 | 1 | 825 | AAI11756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3600267 | Chr.4:38809373 - 38862827 on Build GRCh38 | Loss |
|
| esv2672127 | Chr.4:38809351 - 38862791 on Build GRCh38 | Deletion |
|
| nsv594010 | Chr.4:38804398 - 38873181 on Build GRCh38 | Loss |
|
| esv2762427 | Chr.4:38809930 - 38848489 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map