Assay Details
Target Gene Details
Entrez Gene ID: | 79803 |
Gene Name: | HPS6, biogenesis of lysosomal organelles complex 2 subunit 3 |
Gene Aliases: |
BLOC2S3 |
Location: |
Chr.10:102065367-102068038 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPS6 | NM_024747.5 | 1 | 600 | NP_079023.2 |
| AF536238.1 | 1 | 565 | AAO25965.1 | |
| BC011594.1 | 1 | 565 | AAH11594.2 | |
| BC014993.1 | 1 | 577 | AAH14993.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482149 | Chr.10:102065390 - 102068035 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map