Assay Details
Target Gene Details
Entrez Gene ID: | 57509 |
Gene Name: | microtubule associated tumor suppressor 1 |
Gene Aliases: |
ATBP, ATIP, ICIS, MP44, MTSG1 |
Location: |
Chr.8:17643794-17801579 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2764066 | Chr.8:17530064 - 17677066 on Build GRCh38 | Gain+Loss |
|
| esv3616489 | Chr.8:17490020 - 17737549 on Build GRCh38 | Loss |
|
| nsv831247 | Chr.8:17541589 - 17701918 on Build GRCh38 | Gain |
|
| esv3616492 | Chr.8:17598147 - 17735569 on Build GRCh38 | Loss |
|
| nsv831249 | Chr.8:17644116 - 17838769 on Build GRCh38 | Loss |
|
| esv34867 | Chr.8:17488520 - 17725642 on Build GRCh38 | Loss |
|
| esv3891364 | Chr.8:17633635 - 17678837 on Build GRCh38 | Gain |
|
| esv2759600 | Chr.8:17433671 - 17784021 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73668804,rs73668805] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map