Assay Details
Target Gene Details
Entrez Gene ID: | 1454 |
Gene Name: | casein kinase 1 epsilon |
Gene Aliases: |
CKIepsilon, HCKIE |
Location: |
Chr.22:38290691-38318084 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSNK1E | NM_001894.4 | 11 | 2190 | NP_001885.1 |
| NM_152221.2 | 11 | 2340 | NP_689407.1 |
Target Gene Details
Entrez Gene ID: | 102800317 |
Gene Name: | LOC400927-CSNK1E readthrough |
Gene Aliases: |
- |
Location: |
Chr.22:38290691-38398522 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC400927-CSNK1E | NM_001289912.1 | 15 | 2545 | NP_001276841.1 |
More Information
Set Membership: |
|

Genomic Map