Assay Details
Target Gene Details
Entrez Gene ID: | 64855 |
Gene Name: | family with sequence similarity 129 member B |
Gene Aliases: |
C9orf88, MEG-3, MINERVA, OC58, bA356B19.6 |
Location: |
Chr.9:127505338-127579007 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM129B | NM_001035534.2 | 14 | 2631 | NP_001030611.1 |
| NM_022833.3 | 14 | 2843 | NP_073744.2 | |
| XM_005252135.2 | 15 | 2690 | XP_005252192.2 | |
| XM_011518925.1 | 15 | 2862 | XP_011517227.1 | |
| AB210016.1 | 14 | 2581 | BAE06098.1 | |
| AF151783.1 | 14 | 2613 | AAK57556.1 | |
| AF192911.1 | 14 | 2834 | AAQ13825.1 | |
| AK023580.1 | 6 | 1642 | BAB14615.1 | |
| AK025804.1 | 2 | 999 | BAB15240.1 | |
| AK025951.1 | 1 | 435 | ||
| AL137555.1 | 13 | 2451 | CAB70809.1 | |
| BC001979.1 | 7 | 1730 | AAH01979.2 | |
| BC110345.1 | 5 | 1343 | AAI10346.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv466569 | Chr.9:127470144 - 127512011 on Build GRCh38 | Loss |
|
| nsv615368 | Chr.9:127504287 - 127547894 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs199894907] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map