Assay Details
Target Gene Details
Entrez Gene ID: | 11118 |
Gene Name: | butyrophilin subfamily 3 member A2 |
Gene Aliases: |
BT3.2, BTF4, BTN3.2, CD277 |
Location: |
Chr.6:26365159-26451697 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023453 | Chr.6:25853602 - 26528022 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map