Assay Details
Target Gene Details
Entrez Gene ID: | 5094 |
Gene Name: | poly(rC) binding protein 2 |
Gene Aliases: |
HNRNPE2, HNRPE2, hnRNP-E2 |
Location: |
Chr.12:53452102-53481162 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCBP2 | NM_001098620.2 | NP_001092090.1 | ||
| NM_001128911.1 | NP_001122383.1 | |||
| NM_001128912.1 | NP_001122384.1 | |||
| NM_001128913.1 | NP_001122385.1 | |||
| NM_001128914.1 | NP_001122386.1 | |||
| NM_005016.5 | NP_005007.2 | |||
| NM_031989.4 | NP_114366.1 | |||
| AB188306.1 | BAD36897.1 | |||
| AB208825.1 | BAD92062.1 | |||
| AK225357.1 | ||||
| AK292141.1 | ||||
| AK302067.1 | ||||
| BC001155.1 | AAH01155.1 | |||
| BC071942.1 | AAH71942.1 | |||
| BC107688.1 | AAI07689.1 | |||
| BM764315.1 | ||||
| CX164318.1 | ||||
| X78136.1 | CAA55015.1 |
More Information
Set Membership: |
|

Genomic Map