Assay Details
Target Gene Details
Entrez Gene ID: | 203286 |
Gene Name: | ankyrin repeat and sterile alpha motif domain containing 6 |
Gene Aliases: |
ANKRD14, NPHP16, PKDR1, SAMD6 |
Location: |
Chr.9:98732009-98796542 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKS6 | NM_173551.4 | 15 | 7057 | NP_775822.3 |
| XM_005251793.4 | 16 | 3074 | XP_005251850.1 | |
| XM_005251794.4 | 16 | 3071 | XP_005251851.1 | |
| XM_006716998.3 | 16 | 3105 | XP_006717061.1 | |
| XM_006716999.2 | 16 | 4159 | XP_006717062.1 | |
| XM_011518357.2 | 16 | 5156 | XP_011516659.1 | |
| XM_017014445.1 | 16 | 3102 | XP_016869934.1 | |
| XM_017014446.1 | 15 | 5035 | XP_016869935.1 | |
| XM_017014447.1 | 12 | 1868 | XP_016869936.1 | |
| AK074328.1 | 1 | 3167 | ||
| AK094247.1 | 9 | 1472 | BAC04317.1 | |
| AK126614.1 | 15 | 2173 | ||
| AK127702.1 | 15 | 3196 | ||
| CR749467.1 | 15 | 6728 | CAH18298.1 | |
| CR749472.1 | 13 | 6408 | CAH18302.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv614936 | Chr.9:98420416 - 98918098 on Build GRCh38 | Gain |
|
| esv3621214 | Chr.9:98651384 - 99637088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map