Assay Details
Target Gene Details
Entrez Gene ID: | 29950 |
Gene Name: | SERTA domain containing 1 |
Gene Aliases: |
SEI1, TRIP-Br1 |
Location: |
Chr.19:40422501-40426025 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERTAD1 | NM_013376.3 | 2 | 726 | NP_037508.2 |
| AF117959.1 | 2 | 697 | AAF08349.1 | |
| AF366402.1 | 2 | 678 | AAK52831.1 | |
| AK074652.1 | 2 | 726 | ||
| AK222970.1 | 2 | 693 | BAD96690.1 | |
| AK223011.1 | 2 | 693 | BAD96731.1 | |
| BC002670.2 | 2 | 675 | AAH02670.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833831 | Chr.19:40290833 - 40454958 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map