Assay Details
Target Gene Details
Entrez Gene ID: | 7135 |
Gene Name: | troponin I1, slow skeletal type |
Gene Aliases: |
SSTNI, TNN1 |
Location: |
Chr.1:201403767-201421746 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNNI1 | NM_003281.3 | 9 | 2728 | NP_003272.3 |
| AL831820.1 | 9 | 2712 | CAD38534.1 | |
| AL831975.1 | 9 | 2712 | CAH56221.1 | |
| AL832006.2 | 1 | 1446 | ||
| BX510903.2 | 10 | 2437 | CAD91135.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832315 | Chr.1:201364463 - 201525988 on Build GRCh38 | Loss |
|
| nsv470776 | Chr.1:201319827 - 201534604 on Build GRCh38 | Gain |
|
| nsv508687 | Chr.1:201380850 - 201457524 on Build GRCh38 | Deletion |
|
| nsv467961 | Chr.1:201316584 - 201534604 on Build GRCh38 | Gain |
|
| nsv832326 | Chr.1:201375720 - 201475887 on Build GRCh38 | Loss |
|
| nsv947466 | Chr.1:201308515 - 201540498 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78739506] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map