Assay Details
Target Gene Details
Entrez Gene ID: | 7562 |
Gene Name: | zinc finger protein 708 |
Gene Aliases: |
KOX8, ZNF15, ZNF15L1 |
Location: |
Chr.19:21290388-21329439 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF708 | NM_001297560.1 | 4 | 2209 | NP_001284489.1 |
| NM_001297561.1 | 3 | 2000 | NP_001284490.1 | |
| NM_021269.2 | 4 | 2125 | NP_067092.2 | |
| XM_017027201.1 | 4 | 2027 | XP_016882690.1 | |
| XM_017027202.1 | 4 | 2150 | XP_016882691.1 | |
| XM_017027203.1 | 2 | 1930 | XP_016882692.1 | |
| XM_017027204.1 | XP_016882693.1 | |||
| AK128184.1 | 4 | 2084 | ||
| AK131527.1 | 4 | 2125 | BAD18665.1 | |
| AK311130.1 | 1 | 852 | ||
| BX647526.1 | 4 | 2195 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578950 | Chr.19:21206193 - 21294629 on Build GRCh38 | Loss |
|
| esv2718325 | Chr.19:21119097 - 21656664 on Build GRCh38 | Deletion |
|
| esv2751783 | Chr.19:21283020 - 21543535 on Build GRCh38 | Loss |
|
| esv2665276 | Chr.19:21082662 - 21397631 on Build GRCh38 | Deletion |
|
| nsv578809 | Chr.19:20352186 - 21528400 on Build GRCh38 | Gain |
|
| nsv1061327 | Chr.19:21059770 - 22484758 on Build GRCh38 | Gain |
|
| esv3893179 | Chr.19:21236750 - 21304570 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115298258] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map