Assay Details
Target Gene Details
Entrez Gene ID: | 253738 |
Gene Name: | early B-cell factor 3 |
Gene Aliases: |
COE3, EBF-3, O/E-2, OE-2 |
Location: |
Chr.10:129835232-129964274 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBF3 | NM_001005463.2 | 16 | 2017 | NP_001005463.1 |
| XM_005252667.3 | 17 | 2685 | XP_005252724.1 | |
| XM_005252668.3 | 16 | 2599 | XP_005252725.1 | |
| XM_005252669.3 | 16 | 2572 | XP_005252726.1 | |
| XM_006717739.3 | 17 | 2712 | XP_006717802.1 | |
| XM_006717740.3 | 17 | 2709 | XP_006717803.1 | |
| XM_006717742.3 | 17 | 2604 | XP_006717805.1 | |
| XM_006717743.3 | 16 | 2491 | XP_006717806.1 | |
| XM_006717744.3 | 16 | 2415 | XP_006717807.1 | |
| XM_011539574.2 | 15 | 2427 | XP_011537876.1 | |
| XM_011539575.2 | 12 | 1706 | XP_011537877.1 | |
| XM_017016027.1 | 15 | 2302 | XP_016871516.1 | |
| AL713797.1 | 1 | 656 | ||
| AL832828.1 | 16 | 1944 | CAH56225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832023 | Chr.10:129812459 - 130010617 on Build GRCh38 | Gain+Loss |
|
| esv3577850 | Chr.10:128673756 - 129928631 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map