Assay Details
Target Gene Details
Entrez Gene ID: | 6228 |
Gene Name: | ribosomal protein S23 |
Gene Aliases: |
S23 |
Location: |
Chr.5:82273320-82278416 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS23 | NM_001025.4 | 4 | 1204 | NP_001016.1 |
| AK092335.1 | 1 | 655 | ||
| AK291132.1 | ||||
| BM476492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830364 | Chr.5:82192947 - 82361636 on Build GRCh38 | Loss |
|
| nsv520938 | Chr.5:82245419 - 82424900 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map