Assay Details
Target Gene Details
Entrez Gene ID: | 157657 |
Gene Name: | chromosome 8 open reading frame 37 |
Gene Aliases: |
BBS21, CORD16, RP64, smalltalk |
Location: |
Chr.8:95244913-95270602 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8orf37 | NM_177965.3 | 1 | 39 | NP_808880.1 |
| XM_005250799.3 | 1 | 1407 | XP_005250856.2 | |
| AK055162.1 | BAB70866.1 | |||
| DA473498.1 | 1 | 39 | ||
| DB205618.1 | ||||
| GQ240139.1 |
Target Gene Details
Entrez Gene ID: | 100616530 |
Gene Name: | C8orf37 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:95268836-95810143 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8orf37-AS1 | NR_038202.1 | |||
| NR_038204.1 | ||||
| NR_038205.1 | ||||
| NR_038206.1 | ||||
| GU295153.1 | ||||
| GU295155.1 | ||||
| GU295158.1 | ||||
| GU295159.1 | ||||
| GU295160.1 | ||||
| GU295162.1 | ||||
| GU295164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023866 | Chr.8:95261027 - 95294150 on Build GRCh38 | Gain |
|

Genomic Map