Assay Details
Target Gene Details
Entrez Gene ID: | 23593 |
Gene Name: | heme binding protein 2 |
Gene Aliases: |
C6ORF34B, C6orf34, PP23, SOUL |
Location: |
Chr.6:138404199-138415593 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEBP2 | NM_001326380.1 | NP_001313309.1 | ||
| NM_001326381.1 | NP_001313310.1 | |||
| NM_014320.2 | NP_055135.1 | |||
| XM_017010631.1 | XP_016866120.1 | |||
| AF117616.1 | AAD32099.1 | |||
| AF411610.1 | AAL07394.1 | |||
| AK057928.1 | ||||
| AY427823.1 | AAR88624.1 | |||
| BC008205.1 | AAH08205.1 | |||
| BC010290.1 | AAH10290.1 | |||
| BC016806.1 | ||||
| BC016807.1 | ||||
| BC093037.1 | AAH93037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752090 | Chr.6:138395100 - 138705640 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map