Assay Details
Target Gene Details
Entrez Gene ID: | 23369 |
Gene Name: | pumilio RNA binding family member 2 |
Gene Aliases: |
PUMH2, PUML2 |
Location: |
Chr.2:20248692-20352423 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv29979 | Chr.2:19408886 - 20481173 on Build GRCh38 | Loss |
|
| nsv524207 | Chr.2:20248800 - 20303675 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map