Assay Details
Target Gene Details
Entrez Gene ID: | 26000 |
Gene Name: | TBC1 domain family member 10B |
Gene Aliases: |
EPI64B, FP2461 |
Location: |
Chr.16:30357101-30370258 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D10B | NM_015527.3 | 9 | 2610 | NP_056342.3 |
| XM_011545789.1 | 6 | 2352 | XP_011544091.1 | |
| XM_011545790.2 | 8 | 2477 | XP_011544092.1 | |
| AF370370.1 | 9 | 1756 | AAQ15206.1 | |
| AK002101.1 | 3 | 1113 | BAA92086.1 | |
| AK023192.1 | 9 | 1989 | BAB14454.1 | |
| AL117408.1 | 2 | 1807 | CAB55908.1 | |
| AY007150.1 | 1 | 329 | ||
| BC050523.2 | 5 | 1324 | AAH50523.2 | |
| BC063112.1 | 9 | 2034 | AAH63112.1 | |
| BC072453.1 | 9 | 1922 | ||
| BC098419.1 | 9 | 2174 | AAH98419.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758642 | Chr.16:30062634 - 30371959 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map