Assay Details
Target Gene Details
Entrez Gene ID: | 127294 |
Gene Name: | myomesin 3 |
Gene Aliases: |
- |
Location: |
Chr.1:24056041-24112175 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOM3 | NM_152372.3 | 26 | 3404 | NP_689585.3 |
| AK128233.1 | 18 | 2464 | BAC87343.1 | |
| BC067101.1 | 26 | 3414 | AAH67101.1 | |
| BX537911.1 | 25 | 3128 | CAD97895.1 | |
| BX647453.1 | 26 | 3365 | CAH10377.1 | |
| BX647455.1 | 26 | 3404 | CAH10376.1 | |
| BX647644.1 | 26 | 3365 |
More Information
Set Membership: |
|

Genomic Map