Assay Details
Target Gene Details
Entrez Gene ID: | 6554 |
Gene Name: | solute carrier family 10 member 1 |
Gene Aliases: |
NTCP |
Location: |
Chr.14:69775835-69797289 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC10A1 | NM_003049.3 | NP_003040.1 | ||
| AK314028.1 | ||||
| AY544127.1 | AAT11158.1 | |||
| BC069799.1 | AAH69799.1 | |||
| BC069822.1 | AAH69822.1 | |||
| BC074724.2 | AAH74724.1 | |||
| BC126298.1 | ||||
| BC136355.1 | ||||
| JQ814895.1 | ||||
| L21893.1 | AAA36381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv456336 | Chr.14:69615239 - 70107432 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map