Assay Details
Target Gene Details
Entrez Gene ID: | 140576 |
Gene Name: | S100 calcium binding protein A16 |
Gene Aliases: |
AAG13, DT1P1A7, S100F |
Location: |
Chr.1:153606883-153613168 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A16 | NM_001317007.1 | 3 | 1111 | NP_001303936.1 |
| NM_001317008.1 | 3 | 832 | NP_001303937.1 | |
| NM_080388.2 | 3 | 871 | NP_525127.1 | |
| NR_133645.1 | 3 | 897 | ||
| AI673616.1 | 1 | 342 | ||
| AY762098.1 | 3 | 786 | ||
| BC010541.1 | 3 | 741 | AAH10541.1 | |
| BC019099.1 | 3 | 785 | AAH19099.1 | |
| BC095462.1 | 3 | 706 | AAH95462.1 | |
| U23859.1 |
More Information
Set Membership: |
|

Genomic Map