Assay Details
Target Gene Details
Entrez Gene ID: | 23062 |
Gene Name: | golgi associated, gamma adaptin ear containing, ARF binding protein 2 |
Gene Aliases: |
VEAR |
Location: |
Chr.16:23463542-23510494 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GGA2 | NM_015044.4 | 17 | 4113 | NP_055859.1 |
| XM_017023074.1 | 18 | 7802 | XP_016878563.1 | |
| XM_017023075.1 | 17 | 4116 | XP_016878564.1 | |
| AB029003.1 | 15 | 3713 | BAA83032.1 | |
| BU855556.1 | 1 | 367 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1768 | Chr.16:23450286 - 23482704 on Build GRCh38 | Insertion |
|
| nsv1066078 | Chr.16:23388873 - 23497409 on Build GRCh38 | Gain |
|
| dgv2826n100 | Chr.16:23335983 - 23544166 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map