Assay Details
Target Gene Details
Entrez Gene ID: | 9388 |
Gene Name: | lipase G, endothelial type |
Gene Aliases: |
EDL, EL, PRO719 |
Location: |
Chr.18:49560699-49592908 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPG | NM_001308006.1 | 9 | 1641 | NP_001294935.1 |
| NM_006033.3 | 10 | 1863 | NP_006024.1 | |
| XM_005258390.1 | 10 | 2680 | XP_005258447.1 | |
| XM_011526265.2 | 9 | 1671 | XP_011524567.1 | |
| XM_011526267.1 | 10 | 2250 | XP_011524569.1 | |
| XM_017026095.1 | 10 | 2362 | XP_016881584.1 | |
| AF118767.1 | 10 | 1837 | AAD30434.1 | |
| AK124636.1 | 1 | 1246 | ||
| AK291799.1 | 10 | 1837 | ||
| AK300333.1 | 9 | 1612 | ||
| BC060825.1 | 10 | 1834 | AAH60825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv438288 | Chr.18:49575632 - 49591539 on Build GRCh38 | Loss |
|
| nsv576863 | Chr.18:49569117 - 49938639 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map