Assay Details
Target Gene Details
Entrez Gene ID: | 57586 |
Gene Name: | synaptotagmin 13 |
Gene Aliases: |
- |
Location: |
Chr.11:45240302-45286333 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYT13 | NM_001247987.1 | 8 | 2909 | NP_001234916.1 |
| NM_020826.2 | 6 | 2691 | NP_065877.1 | |
| AB037848.1 | 6 | 2677 | ||
| AL512743.1 | 6 | 2614 | CAC21669.1 |
More Information
Set Membership: |
|

Genomic Map