Assay Details
Target Gene Details
Entrez Gene ID: | 79324 |
Gene Name: | olfactory receptor family 51 subfamily G member 1 (gene/pseudogene) |
Gene Aliases: |
OR11-29, OR51G3P |
Location: |
Chr.11:4923374-4924339 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR51G1 | NM_001005237.1 | 1 | 532 | NP_001005237.1 |
| BC136852.1 | 1 | 600 | ||
| BC136861.1 | 1 | 600 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891914 | Chr.11:4642313 - 4974456 on Build GRCh38 | Gain |
|
| nsv1054847 | Chr.11:4621645 - 5179426 on Build GRCh38 | Gain |
|
| esv2759799 | Chr.11:4886663 - 5114101 on Build GRCh38 | Loss |
|
| dgv306e212 | Chr.11:4916622 - 4924872 on Build GRCh38 | Loss |
|
| esv3579254 | Chr.11:4921674 - 4924872 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map