Assay Details
Target Gene Details
Entrez Gene ID: | 4939 |
Gene Name: | 2'-5'-oligoadenylate synthetase 2 |
Gene Aliases: |
- |
Location: |
Chr.12:112978395-113011723 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OAS2 | NM_002535.2 | 10 | 2781 | NP_002526.2 |
| NM_016817.2 | NP_058197.2 | |||
| BC049215.1 | 10 | 2682 | AAH49215.1 | |
| M87284.1 | 10 | 2660 | AAA60606.1 | |
| M87434.1 | AAA60607.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
| nsv1042636 | Chr.12:112627854 - 113014974 on Build GRCh38 | Gain |
|
| nsv832516 | Chr.12:112883300 - 113040797 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map