Assay Details
Target Gene Details
Entrez Gene ID: | 144455 |
Gene Name: | E2F transcription factor 7 |
Gene Aliases: |
- |
Location: |
Chr.12:77021246-77065580 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2F7 | NM_203394.2 | 13 | 3936 | NP_976328.2 |
| XM_011537966.2 | 12 | 3801 | XP_011536268.1 | |
| XM_011537969.2 | 12 | 3499 | XP_011536271.1 | |
| AK096316.1 | 5 | 2145 | ||
| BC016658.1 | 3 | 1495 | AAH16658.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115820700] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map