Assay Details
Target Gene Details
Entrez Gene ID: | 7567 |
Gene Name: | zinc finger protein 19 |
Gene Aliases: |
KOX12 |
Location: |
Chr.16:71474073-71489351 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF19 | NM_006961.3 | 6 | 1516 | NP_008892.2 |
| AK055567.1 | 5 | 1397 | BAB70957.1 | |
| AK057330.1 | 5 | 1691 | BAB71430.1 | |
| AK292260.1 | 6 | 1714 | ||
| AL834237.1 | 1 | 890 | CAD38915.1 | |
| BC047091.1 | 6 | 1503 | AAH47091.1 |
More Information
Set Membership: |
|

Genomic Map