Assay Details
Target Gene Details
Entrez Gene ID: | 55057 |
Gene Name: | absent in melanoma 1-like |
Gene Aliases: |
CRYBG2 |
Location: |
Chr.1:26321859-26354130 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIM1L | NM_001039775.3 | 17 | 4670 | NP_001034864.2 |
| XM_005245918.2 | 17 | 4656 | XP_005245975.1 | |
| XM_011541672.1 | 16 | 4620 | XP_011539974.1 | |
| XM_011541673.2 | 17 | 4895 | XP_011539975.1 | |
| AK000902.1 | BAA91416.1 | |||
| AK095339.1 | 16 | 1714 | BAC04533.1 | |
| BC136870.1 | 13 | 1412 |
More Information
Set Membership: |
|

Genomic Map