Assay Details
Target Gene Details
Entrez Gene ID: | 10894 |
Gene Name: | lymphatic vessel endothelial hyaluronan receptor 1 |
Gene Aliases: |
CRSBP-1, HAR, LYVE-1, XLKD1 |
Location: |
Chr.11:10557866-10568818 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYVE1 | NM_006691.3 | 1 | 151 | NP_006682.2 |
| AF127670.2 | AAD49220.2 | |||
| AK075443.1 | 1 | 65 | ||
| AK312462.1 | 1 | 148 | ||
| AY358925.1 | AAQ89284.1 | |||
| DA924658.1 | 1 | 151 |
Target Gene Details
Entrez Gene ID: | 100129827 |
Gene Name: | MRVI1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:10541236-10599932 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRVI1-AS1 | NR_034093.2 | |||
| NR_034094.2 | ||||
| NR_046374.1 | ||||
| NR_046375.1 | ||||
| AI075905.1 | ||||
| BC028123.1 | AAH28123.1 | |||
| BI520309.1 | ||||
| BU569149.1 | ||||
| BU570065.1 | ||||
| DB071465.1 | ||||
| DB454301.1 |
More Information
Set Membership: |
|

Genomic Map