Assay Details
Target Gene Details
Entrez Gene ID: | 57403 |
Gene Name: | RAB22A, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.20:58309715-58367507 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB22A | NM_020673.2 | NP_065724.1 | ||
| AF091034.2 | AAF00047.2 | |||
| AJ276210.1 | CAC10538.1 | |||
| AK091180.1 | ||||
| BC015710.1 | AAH15710.1 | |||
| BC063457.1 | AAH63457.1 | |||
| BI457862.1 | ||||
| BT007046.1 | AAP35695.1 | |||
| DB452461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055956 | Chr.20:58211957 - 58367256 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map