Assay Details
Target Gene Details
Entrez Gene ID: | 1944 |
Gene Name: | ephrin A3 |
Gene Aliases: |
EFL2, EPLG3, Ehk1-L, LERK3 |
Location: |
Chr.1:155078872-155087538 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFNA3 | NM_004952.4 | NP_004943.1 | ||
| AK296030.1 | ||||
| AK316248.1 | ||||
| BC017722.1 | AAH17722.1 | |||
| BC110406.1 | ||||
| L37360.1 | AAA52368.1 | |||
| U14187.1 | AAC50078.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| esv3587616 | Chr.1:155055926 - 155098657 on Build GRCh38 | Gain |
|
| esv3587615 | Chr.1:155047853 - 155175013 on Build GRCh38 | Gain |
|
| nsv952212 | Chr.1:154998225 - 155087624 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77832800] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map