Assay Details
Target Gene Details
Entrez Gene ID: | 200909 |
Gene Name: | 5-hydroxytryptamine receptor 3D |
Gene Aliases: |
5HT3D |
Location: |
Chr.3:184031544-184039369 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR3D | NM_182537.2 | NP_872343.2 | ||
| AY159812.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4145 | Chr.3:184023744 - 184056957 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map