Assay Details
Target Gene Details
Entrez Gene ID: | 282763 |
Gene Name: | olfactory receptor family 51 subfamily B member 5 |
Gene Aliases: |
HOR5'Beta5, OR11-37 |
Location: |
Chr.11:5340883-5505652 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR51B5 | NM_001005567.2 | NP_001005567.2 | ||
| NR_038321.1 | ||||
| BC022184.1 | ||||
| BG493214.1 | ||||
| BG776801.1 |
Target Gene Details
Entrez Gene ID: | 390059 |
Gene Name: | olfactory receptor family 51 subfamily M member 1 |
Gene Aliases: |
HOR5'Beta7, OR11-40 |
Location: |
Chr.11:5389377-5390434 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR51M1 | NM_001004756.2 | 1 | 44 | NP_001004756.2 |
| BC136912.1 | 1 | 44 | ||
| BC136913.1 | 1 | 44 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv22160 | Chr.11:5388564 - 5391379 on Build GRCh38 | Loss |
|
| nsv832057 | Chr.11:5354355 - 5554970 on Build GRCh38 | Loss |
|
| nsv1044243 | Chr.11:5379806 - 5411086 on Build GRCh38 | Loss |
|
| esv2761646 | Chr.11:5384893 - 5433011 on Build GRCh38 | Loss |
|
| nsv1046068 | Chr.11:5314713 - 6042512 on Build GRCh38 | Gain |
|
| nsv1048608 | Chr.11:5307286 - 5623949 on Build GRCh38 | Gain |
|
| nsv524400 | Chr.11:5307131 - 5395392 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116535306] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map