Assay Details
Target Gene Details
Entrez Gene ID: | 2145 |
Gene Name: | enhancer of zeste 1 polycomb repressive complex 2 subunit |
Gene Aliases: |
KMT6B |
Location: |
Chr.17:42700275-42745096 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EZH1 | NM_001321079.1 | 20 | 3006 | NP_001308008.1 |
| NM_001321081.1 | 21 | 3070 | NP_001308010.1 | |
| NM_001321082.1 | 20 | 2977 | NP_001308011.1 | |
| NM_001991.4 | 21 | 3097 | NP_001982.2 | |
| XM_005257145.2 | 19 | 2852 | XP_005257202.1 | |
| XM_011524517.2 | 20 | 2945 | XP_011522819.1 | |
| XM_017024350.1 | 20 | 2924 | XP_016879839.1 | |
| XM_017024351.1 | 19 | 2831 | XP_016879840.1 | |
| XM_017024352.1 | 18 | 2682 | XP_016879841.1 | |
| AB002386.1 | 20 | 3009 | ||
| AL833159.1 | 21 | 3017 | ||
| BC015882.1 | 21 | 3031 | AAH15882.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833450 | Chr.17:42660985 - 42825144 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73983746] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map