Assay Details
Target Gene Details
| Entrez Gene ID: | 4852 | 
| Gene Name: | neuropeptide Y | 
| Gene Aliases: | PYY4 | 
| Location: | Chr.7:24284188-24291865 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Exon 2 - Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| NPY | NM_000905.3 | NP_000896.1 | ||
| AI198311.1 | ||||
| AK312156.1 | ||||
| BC029497.1 | AAH29497.1 | |||
| K01911.1 | AAA59944.1 | |||
| M15789.1 | AAA59946.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2673027 | Chr.7:23679603 - 24432457 on Build GRCh38 | Deletion |  FAM221A  STK31  LOC107986777  NPY | 
| nsv830927 | Chr.7:24262646 - 24438857 on Build GRCh38 | Gain |  LOC107986777  NPY | 
| nsv517053 | Chr.7:24224894 - 24386936 on Build GRCh38 | Gain+Loss |  LOC107986777  NPY | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top