Assay Details
Target Gene Details
Entrez Gene ID: | 220979 |
Gene Name: | chromosome 10 open reading frame 25 |
Gene Aliases: |
- |
Location: |
Chr.10:44993932-45000910 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf25 | NM_001039380.3 | 2 | 2169 | NP_001034469.2 |
| NR_126421.1 | ||||
| XM_017015895.1 | 3 | 1300 | XP_016871384.1 | |
| AK055129.1 | 2 | 2147 | BAB70858.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576560 | Chr.10:44916005 - 45054697 on Build GRCh38 | Gain |
|
| esv3623079 | Chr.10:44993759 - 45028111 on Build GRCh38 | Loss |
|
| nsv831848 | Chr.10:44994728 - 45172222 on Build GRCh38 | Gain |
|
| nsv831846 | Chr.10:44826788 - 45060922 on Build GRCh38 | Loss |
|

Genomic Map