Assay Details
Target Gene Details
Entrez Gene ID: | 55177 |
Gene Name: | regulator of microtubule dynamics 3 |
Gene Aliases: |
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51 |
Location: |
Chr.15:40735884-40755336 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMDN3 | NM_001304802.1 | 3 | 498 | NP_001291731.1 |
| NM_001323894.1 | 3 | 412 | NP_001310823.1 | |
| NM_001323895.1 | 3 | 528 | NP_001310824.1 | |
| NM_001323896.1 | 3 | 412 | NP_001310825.1 | |
| NM_001323897.1 | 3 | 528 | NP_001310826.1 | |
| NM_018145.2 | 3 | 528 | NP_060615.1 | |
| AB000782.1 | 2 | 267 | BAB46923.1 | |
| AK001441.1 | 3 | 452 | BAA91693.1 | |
| AK092058.1 | 3 | 411 | ||
| AK097286.1 | 4 | 340 | ||
| AK123192.1 | BAC85554.1 | |||
| AK123282.1 | 2 | 1335 | ||
| AY358793.1 | 2 | 268 | AAQ89153.1 | |
| BC063844.1 | 3 | 416 | AAH63844.1 | |
| DA280017.1 | 3 | 384 | ||
| DA592879.1 | 3 | 412 |
More Information
Set Membership: |
|

Genomic Map