Assay Details
Target Gene Details
Entrez Gene ID: | 54802 |
Gene Name: | tRNA isopentenyltransferase 1 |
Gene Aliases: |
GRO1, IPPT, IPT, IPTase, MOD5, hGRO1 |
Location: |
Chr.1:39841031-39883511 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |
|
| dgv153n100 | Chr.1:39715819 - 39848248 on Build GRCh38 | Gain |
|
| esv3585792 | Chr.1:39811141 - 39845795 on Build GRCh38 | Loss |
|
| esv2666580 | Chr.1:39809257 - 39898016 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map