Assay Details
Target Gene Details
Entrez Gene ID: | 158358 |
Gene Name: | KIAA2026 |
Gene Aliases: |
- |
Location: |
Chr.9:5860254-6008479 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA2026 | XM_011517759.2 | XP_011516061.1 | ||
| XM_011517760.2 | XP_011516062.1 | |||
| XM_017014335.1 | XP_016869824.1 |
Target Gene Details
Entrez Gene ID: | 2315 |
Gene Name: | melan-A |
Gene Aliases: |
MART-1, MART1 |
Location: |
Chr.9:5890909-5909822 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLANA | NM_005511.1 | 5 | 618 | NP_005502.1 |
| BC014423.1 | AAH14423.1 | |||
| U06452.1 | 5 | 618 | AAA19238.1 | |
| U06654.1 | AAA20389.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6464 | Chr.9:5883378 - 5914698 on Build GRCh38 | Insertion |
|
| nsv1029665 | Chr.9:5687332 - 6775107 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map