Assay Details
Target Gene Details
Entrez Gene ID: | 8808 |
Gene Name: | interleukin 1 receptor like 2 |
Gene Aliases: |
IL-1Rrp2, IL-36R, IL1R-rp2, IL1RRP2 |
Location: |
Chr.2:102186973-102242906 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1RL2 | NM_003854.2 | NP_003845.2 | ||
| XM_005264035.1 | XP_005264092.1 | |||
| XM_006712820.2 | XP_006712883.1 | |||
| XM_011512091.1 | XP_011510393.1 | |||
| XM_011512092.1 | XP_011510394.1 | |||
| XM_011512093.1 | XP_011510395.1 | |||
| XM_011512096.1 | XP_011510398.1 | |||
| XM_017005174.1 | XP_016860663.1 | |||
| XM_017005175.1 | XP_016860664.1 | |||
| XM_017005176.1 | XP_016860665.1 | |||
| XM_017005177.1 | XP_016860666.1 | |||
| XM_017005178.1 | XP_016860667.1 | |||
| XM_017005179.1 | XP_016860668.1 | |||
| AF284434.1 | AAG21368.1 | |||
| BC107064.2 | ||||
| U49065.1 | AAB53237.1 |
More Information
Set Membership: |
|

Genomic Map