Assay Details
Target Gene Details
Entrez Gene ID: | 3848 |
Gene Name: | keratin 1 |
Gene Aliases: |
CK1, EHK, EHK1, EPPK, K1, KRT1A, NEPPK |
Location: |
Chr.12:52674736-52680407 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT1 | NM_006121.3 | 7 | 1356 | NP_006112.3 |
| AK313986.1 | 7 | 1356 | ||
| BC063697.1 | 7 | 1338 | AAH63697.1 | |
| M10938.1 | 7 | 844 | AAA36153.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428281 | Chr.12:52570654 - 52761443 on Build GRCh38 | Gain |
|
| nsv832415 | Chr.12:52567563 - 52757300 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map