Assay Details
Target Gene Details
Entrez Gene ID: | 79807 |
Gene Name: | glutathione S-transferase C-terminal domain containing |
Gene Aliases: |
- |
Location: |
Chr.4:105708778-105848023 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTCD | NM_001031720.3 | 3 | 541 | NP_001026890.2 |
| NM_024751.3 | 3 | 462 | NP_079027.2 | |
| XM_005263222.3 | 3 | 729 | XP_005263279.1 | |
| XM_011532248.2 | 3 | 685 | XP_011530550.1 | |
| XM_011532249.2 | 3 | 547 | XP_011530551.1 | |
| XM_011532252.2 | 3 | 919 | XP_011530554.1 | |
| AK023335.1 | 3 | 720 | BAB14532.1 | |
| AK225563.1 | 3 | 720 | ||
| AK292355.1 | 3 | 544 | ||
| AK303547.1 | ||||
| BC032942.2 | 3 | 541 | AAH32942.1 | |
| BX648355.1 | 3 | 463 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000196 | Chr.4:103475625 - 106148054 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map