Assay Details
Target Gene Details
Entrez Gene ID: | 7704 |
Gene Name: | zinc finger and BTB domain containing 16 |
Gene Aliases: |
PLZF, ZNF145 |
Location: |
Chr.11:114059576-114256765 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB16 | NM_001018011.1 | 2 | 274 | NP_001018011.1 |
| NM_006006.4 | 2 | 429 | NP_005997.2 | |
| XM_005271658.4 | 2 | 561 | XP_005271715.1 | |
| XM_017018257.1 | 2 | 362 | XP_016873746.1 | |
| XM_017018258.1 | 2 | 517 | XP_016873747.1 | |
| XM_017018259.1 | 2 | 869 | XP_016873748.1 | |
| AB208916.1 | 2 | 413 | BAD92153.1 | |
| BC026902.1 | 2 | 333 | AAH26902.1 | |
| BC029812.1 | 2 | 411 | AAH29812.1 | |
| BX648973.1 | 2 | 546 | ||
| KU178400.1 | 1 | 165 | ||
| KU178401.1 | 1 | 165 | ||
| Z19002.1 | 1 | 240 | CAA79489.1 |
More Information
Set Membership: |
|

Genomic Map