Assay Details
Target Gene Details
Entrez Gene ID: | 2542 |
Gene Name: | solute carrier family 37 member 4 |
Gene Aliases: |
G6PT1, G6PT2, G6PT3, GSD1b, GSD1c, GSD1d, PRO0685, TRG-19, TRG19 |
Location: |
Chr.11:119024351-119030906 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC37A4 | NM_001164277.1 | 3 | 716 | NP_001157749.1 |
| NM_001164278.1 | 3 | 400 | NP_001157750.1 | |
| NM_001164279.1 | NP_001157751.1 | |||
| NM_001164280.1 | 1 | 158 | NP_001157752.1 | |
| NM_001467.5 | 2 | 212 | NP_001458.1 | |
| AB209026.1 | 1 | 1453 | BAD92263.1 | |
| AK289642.1 | 3 | 684 | ||
| AK294993.1 | 2 | 178 | ||
| AK298395.1 | ||||
| AK300646.1 | ||||
| AK312628.1 | 3 | 382 | ||
| AY423732.1 | AAS00495.1 | |||
| BC002400.2 | 1 | 160 | AAH02400.1 | |
| BC003589.1 | 1 | 160 | AAH03589.1 | |
| BC014663.1 | 1 | 167 | AAH14663.1 | |
| BC015650.2 | 2 | 632 | AAH15650.1 | |
| BC064563.1 | 1 | 155 | AAH64563.1 | |
| DA485567.1 | 2 | 186 | ||
| DC295438.1 | 2 | 212 | ||
| Y15409.1 | 1 | 128 | CAA75608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952744 | Chr.11:119024091 - 119030790 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map