Assay Details
Target Gene Details
Entrez Gene ID: | 4184 |
Gene Name: | sperm mitochondria associated cysteine rich protein |
Gene Aliases: |
HSMCSGEN1, MCS, MCSP |
Location: |
Chr.1:152878322-152885047 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMCP | NM_030663.2 | NP_109588.2 | ||
| AK312020.1 | ||||
| BC014593.2 | AAH14593.1 | |||
| BC016744.2 | AAH16744.1 | |||
| X89960.1 | CAA62000.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv432n100 | Chr.1:152777257 - 152889390 on Build GRCh38 | Gain |
|
| nsv831592 | Chr.1:152842578 - 153024686 on Build GRCh38 | Gain |
|
| nsv547899 | Chr.1:152699529 - 153272606 on Build GRCh38 | Gain |
|
| esv2758970 | Chr.1:152500363 - 152925677 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map