Assay Details
Target Gene Details
Entrez Gene ID: | 1519 |
Gene Name: | cathepsin O |
Gene Aliases: |
CTSO1 |
Location: |
Chr.4:155924118-155953896 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSO | NM_001334.2 | NP_001325.1 | ||
| AK300112.1 | ||||
| AK308323.1 | ||||
| AK312354.1 | ||||
| BC049206.1 | AAH49206.1 | |||
| BI834113.1 | ||||
| X77383.1 | CAA54562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575777 | Chr.4:155855410 - 156699594 on Build GRCh38 | Gain |
|
| nsv830121 | Chr.4:155850003 - 156002314 on Build GRCh38 | Gain |
|
| esv3569758 | Chr.4:155935481 - 155943599 on Build GRCh38 | Loss |
|
| nsv1029122 | Chr.4:155773161 - 156031641 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map